(4-Bromophenyl)acetone 97%

Synonyms: 4-Bromophenylacetone

AC38022-0050 AC38022-0250
200052-128EA 75.4 USD
200052-128 103513-072
(4-Bromophenyl)acetone 97%
(4-Bromophenyl)acetone
Formula: C₉H₉BrO
MW: 213.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210401
CAS Number: 6186-22-7

Order Now

Specification Test Results

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5560 to 1.5590 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR