6-Bromopicolinic acid 98%

Warning

Synonyms: 6-Bromo-2-pyridinecarboxylic acid6-Bromopyridine-2-carboxylic acid

AC37993-0050 AC37993-0010
200054-934EA 151 USD
200054-934 200054-932
6-Bromopicolinic acid 98%
6-Bromopicolinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 192…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00093197
CAS Number: 21190-87-4

Order Now

Specification Test Results

Appearance Off-white to tan Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR