Ethyl-4-oxocyclohexanecarboxylate 97%

Warning

Synonyms: Ethyl 4-oxocyclohexanecarboxylate

AC37991-0010 AC37991-0050 AC37991-2500
200055-566EA 25 USD
200055-566 200055-568 103513-066
Ethyl-4-oxocyclohexanecarboxylate 97%
Ethyl 4-oxocyclohexanecarboxylate

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4610 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR