2,5-Dichloro-3-nitropyridine 97%
|
Warning
|
AC37952-0010
AC37952-0050
200053-960EA
72.4
USD
200053-960
200053-958
2,5-Dichloro-3-nitropyridine 97%
2,5-Dichloro-3-nitropyridine
Specification Test Results
Appearance | Green-beige to orange Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 39 to 45 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...