5-Allyl-3-methoxysalicylaldehyde 98%

Synonyms: 5-Allyl-2-hydroxy-3-methoxybenzaldehyde

AC37936-0100 AC37936-0250
200054-808EA 80.1 USD
200054-808 200054-810
5-Allyl-3-methoxysalicylaldehyde 98%
5-Allyl-3-methoxysalicylaldehyde
Formula: C₁₁H₁₂O₃
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD06408022
CAS Number: 22934-51-6

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 48 to 52 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR