(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane 90%

Warning

Synonyms: (R,R)-Ethylenebis[(2-methoxyphenyl)phenylphosphine](R,R)-DIPAMP(R,R)-(-)-1,2-Bis[(2-methoxyphenyl)(phenyl)phosphino]ethane(1R,2R)-Bis[(2-methoxyphenyl)phenylphosphino]ethane(-)-DIPAMP

AC37935-0010
200053-290EA 314 USD
200053-290
(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane 90%
(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane

Appearance: White Crystalline powder

Formula: C₂₈H₂₈O₂P₂
MW: 458.48 g/mol
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD05863546
CAS Number: 55739-58-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Specific optical rotation -80° to -85° (20 °C, 589 nm) (C=1, CHCl3)
Titration with HClO4 ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR