(2,6-Difluorophenyl)acetone 98%
AC37915-0250
AC37915-0010
AC37915-0050
200053-996EA
192
USD
200053-996
103513-062
103513-060
(2,6-Difluorophenyl)acetone 98%
(2,6-Difluorophenyl)acetone
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4790 to 1.4820 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...