Dibutylboron trifluoromethanesulfonate 1 M in dichloromethane

Danger

AC37899-1000 AC37899-0250
200055-426EA 233.08 USD
200055-426 103528-112
Dibutylboron trifluoromethanesulfonate 1 M in dichloromethane
Dibutylboron trifluoromethanesulfonate
Formula: C₉H₁₈BF₃O₃S
Density: 1.271 g/cm³
Flash Pt: 27 °C
Storage Temperature: Freezer
MDL Number: MFCD00009669
CAS Number: 60669-69-4
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to orange Solution
Strength 0.9 to 1.1 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR