4-Methoxy-3-pyridineboronic acid hydrate 97%

Warning

Synonyms: 4-Methoxypyridine-3-boronic acid hydrate

AC37838-0010 AC37838-2500
103513-046EA 471 USD
103513-046 200054-714
4-Methoxy-3-pyridineboronic acid hydrate 97%
4-Methoxy-3-pyridineboronic acid hydrate
Formula: C₆H₈BNO₃·H₂O
MW: 152.94 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD07186213
CAS Number: 1266615-79-5

Order Now

Specification Test Results

Appearance Cream Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR