3,6-Dimethyl-1,4-dioxane-2,5-dione 99%

Warning

Synonyms: DL-Lactide

AC37685-0250 AC37685-1000
200054-352EA 69.6 USD
200054-352 200054-350
3,6-Dimethyl-1,4-dioxane-2,5-dione 99%
3,6-Dimethyl-1,4-dioxane-2,5-dione
Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 138…142 °C (8 mmHg)
Melting Pt: 122…126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011685
CAS Number: 95-96-5
EINECS: 202-468-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 122 to 126 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR