Dimethyl-L-aspartate hydrochloride 98%

Synonyms: Dimethyl L-Asparaginate HydrochlorideL-Asparaginic Acid Dimethyl Ester Hydrochloride(S)-Aminosuccinic Acid Dimethyl Ester HydrochlorideH-Asp(ome)-OMe HClL-Aspartic Acid Dimethyl Ester HydrochlorideDimethyl (S)-Aminosuccinate HydrochlorideH-Asp(OMe)-OMe.HCl

AC37666-0050 AC37666-0250
200052-738EA 62.4 USD
200052-738 200052-736
Dimethyl-L-aspartate hydrochloride 98%
Dimethyl-L-aspartate hydrochloride

Appearance: White to off-white Crystalline powder

Formula: C₆H₁₁NO₄
MW: 197.62 g/mol
Melting Pt: 115…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00038878
CAS Number: 32213-95-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Specific optical rotation +13° to +17° (20 °C, 589 nm) (c=1, CH3OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR