4-(Morpholin-4-ylmethyl)benzonitrile 97%

Danger

Synonyms: 4-(Morpholinomethyl)benzonitrile

AC37633-0010
200052-040EA 44 USD
200052-040
4-(Morpholin-4-ylmethyl)benzonitrile 97%
4-(Morpholin-4-ylmethyl)benzonitrile
Formula: C₁₂H₁₄N₂O
MW: 202.2500 g/mol
Melting Pt: 84…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00454271
CAS Number: 37812-51-4
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 84 to 89 °C
GC ≥96 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR