3,5-Dichloro-1,2,4-thiadiazole 97%
|
Warning
|
AC37603-0250
AC37603-0010
AC37603-0050
103512-960EA
665
USD
103512-960
200023-074
200051-942
3,5-Dichloro-1,2,4-thiadiazole 97%
3,5-Dichloro-1,2,4-thiadiazole
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5570 to 1.5600 (20 °C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...