4,4'-Dimethyl-2,2'-bipyridyl 99+%

Warning

Synonyms: 4,4'-Dimethyl-2,2'-bipyridine

AC37323-0250 AC37323-0050 AC37323-0010
200054-540EA 123 USD
200054-540 103528-008 200054-538
4,4'-Dimethyl-2,2'-bipyridyl 99+%
4,4'-Dimethyl-2,2'-bipyridyl
Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00006441
CAS Number: 1134-35-6
EINECS: 214-483-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige Crystalline powder
Infrared spectrum Conforms
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR