Cyclopentyl isocyanide 96%

Warning

AC37209-0050 AC37209-0010
103532-672EA 594 USD
103532-672 200063-794
Cyclopentyl isocyanide 96%
Cyclopentyl isocyanide
Formula: C₆H₉N
Boiling Pt: 40 °C (11hPa)
Density: 0.86 g/cm³
Storage Temperature: Freezer
MDL Number: MFCD02664624
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4410 (20 °C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR