Ethylboronic acid 95%

Warning

AC37086-0050 AC37086-0010
200012-464EA 40.3 USD
200012-464 103512-906
Ethylboronic acid 95%
Ethylboronic acid
Formula: C₂H₇BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01074536
CAS Number: 4433-63-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or flakes or needles
Infrared spectrum Conforms
Titration with NaOH ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR