Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate 90%

Warning

Synonyms: Methyl [Bis(2,2,2-trifluoroethoxy)phosphinyl]acetateBis(2,2,2-trifluoroethyl)phosphonoacetic Acid Methyl EsterMethyl Bis(2,2,2-trifluoroethyl)phosphonoacetateMethyl O,O'-bis(2,2,2-trifluoroethyl)phosphonoacetateMethyl 2-(bis(2,2,2-trifluoroethoxy)phosphoryl)acetate

AC37061-0050
200013-308EA 162 USD
200013-308
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate 90%
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate

Appearance: Clear orange to dark brown Liquid

Formula: C₇H₉F₆O₅P
MW: 318.11 g/mol
Density: 1.504 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009901
CAS Number: 88738-78-7

Order Now

Specification Test Results

Appearance Clear orange to dark brown Liquid
Infrared spectrum Conforms
Refractive index 1.3700 to 1.3740 (20 °C, 589 nm)
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR