Acetoxyacetyl chloride 97%

Danger

Synonyms: Acetoxyacetylchloride2-Chloro-2-oxoethyl acetate

AC37007-0050 AC37007-0250
200017-986EA 86.9 USD
200017-986 200024-292
Acetoxyacetyl chloride 97%
Acetoxyacetyl chloride
Formula: C₄H₅ClO₃
MW: 136.53 g/mol
Boiling Pt: 64…68 °C (25 mmHg)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00011535
CAS Number: 13831-31-7
EINECS: 237-542-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4255 to 1.4275 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR