Isosorbide dimethyl ether 99%

Synonyms: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan

AC36991-1000 AC36991-0250
200012-894EA 184 USD
200012-894 200031-636
Isosorbide dimethyl ether 99%
Isosorbide dimethyl ether
Formula: C₈H₁₄O₄
MW: 174.20 g/mol
Boiling Pt: 93…95 °C (.1hPa)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00064828
CAS Number: 5306-85-4
EINECS: 226-159-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4620 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR