3-(Methoxycarbonyl)phenylboronic acid ≥97%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: 3-(Methoxycarbonyl)benzeneboronic acid

AC36983-0050 AC36983-0250 AC36983-0010
200005-264EA 147.51 USD
200005-264 103512-890 200023-360
3-(Methoxycarbonyl)phenylboronic acid ≥97%
3-(Methoxycarbonyl)phenylboronic acid
Formula: C₈H₉BO₄
MW: 179,97
Melting Pt: 187…191 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093046
CAS Number: 99769-19-4
EINECS: 000-000-0
UN: 0000

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR