2-Bromoterephthalic acid 95%

Warning

AC36981-0250 AC36981-1000
200034-668EA 217 USD
200034-668 103464-420
2-Bromoterephthalic acid 95%
2-Bromoterephthalic acid
Formula: C₈H₅BrO₄
Melting Pt: 295…297 °C
Storage Temperature: Ambient
MDL Number: MFCD00002403
CAS Number: 586-35-6
EINECS: 209-572-8

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Titration with NaOH ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR