Amberlite® IR-120 (H) ion-exchange resin

Danger

AC36972-2500
103467-288EA 43.38 USD
103467-288
Amberlite® IR-120 (H) ion-exchange resin
Amberlite® IR-120 (H)
Storage Temperature: Ambient
MDL Number: MFCD00132707
CAS Number: 78922-04-0

Order Now

Specification Test Results

Appearance Amber to yellow-brown Beads
Particle size ≤2.0 % (< 0.300 mm)
Particle size ≤3.0 % (> 1.180 mm)
Harmonic mean size 0.620 to 0.830 mm
Uniformity coefficient ≤1.8
Moisture holding capacity 53 to 58 % (H+)
Total volume capacity ≥1.8 eq/l (H+)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR