Bis(pyridine)iodonium tetrafluoroborate 97%

Warning

AC36968-0010
200005-866EA 63.1 USD
200005-866
Bis(pyridine)iodonium tetrafluoroborate 97%
Bis(pyridine)iodonium tetrafluoroborate
Formula: C₁₀H₁₀BF₄IN₂
Melting Pt: 137…141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03703393
CAS Number: 15656-28-7

Order Now

Specification Test Results

Appearance Light yellow to orange to tan Powder or crystals
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR