Methyl thiosalicylate 97%

Warning

Synonyms: Methyl 2-mercaptobenzoate

AC36960-0050 AC36960-0250
200025-894EA 80.9 USD
200025-894 200043-940
Methyl thiosalicylate 97%
Methyl thiosalicylate
Formula: C₈H₈O₂S
MW: 168.21 g/mol
Boiling Pt: 98…100 °C (2 mmHg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00060678
CAS Number: 4892-02-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light green Liquid
Infrared spectrum Conforms
Refractive index 1.5910 to 1.5950 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR