1-Methylimidazole-2-carbaldehyde 97%

Warning

Synonyms: 1-Methyl-1H-imidazole-2-carbaldehyde1-Methylimidazole-2-carboxaldehyde

AC36985-0050 AC36985-0010
200004-764EA 45.4 USD
200004-764 200008-710
1-Methylimidazole-2-carbaldehyde 97%
1-Methylimidazole-2-carbaldehyde
Formula: C₅H₆N₂O
MW: 110.12 g/mol
Boiling Pt: 70…74 °C (1 mmHg)
Melting Pt: 35…39 °C
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD01321308
CAS Number: 13750-81-7

Order Now

Specification Test Results

Appearance White to yellow to orange Crystals or needles or crystalline mass and/or chunks
Infrared spectrum Conforms
Melting point 35 to 39 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR