(S)-(-)-Tetrahydro-2-furoic acid 97%, 98% ee

Danger

Synonyms: (2S)-(-)-Tetrahydrofuran-2-carboxylic acid(S)-Tetrahydrofuran-2-carboxylic acid

AC36951-0010
200038-740EA 30.4 USD
200038-740
(S)-(-)-Tetrahydro-2-furoic acid 97%, 98% ee
(S)-(-)-Tetrahydro-2-furoic acid
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 244 °C (736 mmHg)
Density: 1.21 g/cm³
Flash Pt: 139 °C
Storage Temperature: Ambient
MDL Number: MFCD00272639
CAS Number: 87392-07-2
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20 °C, 589 nm)
Enantiomeric excess ≥97.5 %
Specific optical rotation -33° to -39° (20 °C, 589 nm) (c=1, CHCl3)
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR