Bis(2-methoxyethyl)aminosulfur trifluoride 50% (w/w) 2.7 M in toluene

Danger

AC36896-0050 AC36896-0250 AC36896-1000
103524-278EA 695.77 USD
103524-278 200056-734 200056-732
Bis(2-methoxyethyl)aminosulfur trifluoride 50% (w/w) 2.7 M in toluene
Bis(2-methoxyethyl)aminosulfur trifluoride
Formula: C₆H₁₄F₃NO₂S
MW: 221.24 g/mol
Density: 1.2 g/cm³
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01321415
CAS Number: 202289-38-1
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance Dark yellow to orange Liquid
NMR ≥95 % (on neat substance)
Titration with HClO4 ≥50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR