(4-Methoxyphenyl)acetyl chloride 98%
|
Danger
|
Synonyms:
4-Methoxyphenylacetyl chloride
AC36893-0100
AC36893-0020
200029-882EA
26.4
USD
200029-882
200035-968
(4-Methoxyphenyl)acetyl chloride 98%
(4-Methoxyphenyl)acetyl chloride
Formula:
C₉H₉ClO₂ MW: 184.62 g/mol Boiling Pt: 143 °C (10 mmHg) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075474 CAS Number: 4693-91-8 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow to red Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5390 to 1.5420 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...