Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aminoguanidine hydrochloride ≥98%

Supplier: ACROS ORGANICS MS
Warning

AC36891-0250 AC36891-1000
200011-184 200011-182
Aminoguanidine hydrochloride ≥98%


Formula: CH₇ClN₄
MW: 110,55 g/mol
Storage Temperature: Ambient
CAS Number: 1937-19-5
EINECS: 217-707-7
Order Now
Specification Test Results
Appearance White to off-white Crystals
Infrared spectrum Conforms
Melting point ≥160°C
Titration with HClO4 ≥97.5 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty