2-Aminoimidazole hemisulfate 98+%

Warning

Synonyms: 2-Aminoimidazole sulphate2-Aminoimidazole sulfate1H-imidazol-2-amine sulfate(2:1)1H-imidazol-2-amine sulphate(2:1)

AC36886-0100 AC36886-0025
200034-622EA 280 USD
200034-622 200028-526
2-Aminoimidazole hemisulfate 98+%
2-Aminoimidazole hemisulphate
Formula: C₆H₁₂N₆O₄S
MW: 264,26 g/mol
Melting Pt: 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013162
CAS Number: 1450-93-7
EINECS: 215-918-9

Order Now

Specification Test Results

Appearance Brown Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR