2,2,3,3,4,4,4-Heptafluorobutylamine 97%

Danger

AC36877-0050 AC36877-0010
200022-008EA 179 USD
200022-008 200022-006
2,2,3,3,4,4,4-Heptafluorobutylamine 97%
2,2,3,3,4,4,4-Heptafluorobutylamine
Formula: C₄H₄F₇N
MW: 199.07 g/mol
Storage Temperature: Ambient
EINECS: 206-780-0
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR