Indium(III) trifluoromethanesulfonate 98%

Danger

Synonyms: Indium triflateIndium(III) trifluoromethanesulfonateIndium(III) trifluoromethanesulphonate

AC36854-0050 AC36854-0250
200043-662EA 350 USD
200043-662 200019-364
Indium(III) trifluoromethanesulfonate 98%
Indium (III) trifluoromethanesulphonate
Formula: C₃F₉InO₉S₃
MW: 562.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144478
CAS Number: 128008-30-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to almost white Powder and/or granules
Infrared spectrum Conforms
Titration Complexometric 19.9 to 20.9 % (In)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR