(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 98%

Warning

AC36841-0010 AC36841-0050
200051-194EA 160 USD
200051-194 200015-234
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 98%
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol
Formula: C₁₄H₁₅NO
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00074959
CAS Number: 23364-44-5

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 141 to 145 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR