1-Boc-4-hydroxypiperidine 97%

Warning

Synonyms: N-BOC-4-HydroxypiperidineN-Boc-4-piperidinoltert-butyl 4-hydroxypiperidine-1-carboxylatetert-Butyl 4-hydroxypiperidine-1-carboxylate

AC36827-0050
200013-582EA 51.8 USD
200013-582
1-Boc-4-hydroxypiperidine 97%
1-Boc-4-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 60…65 °C
Storage Temperature: Ambient
MDL Number: MFCD01075174
CAS Number: 109384-19-2

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 60 to 65 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR