3-Bromo-3-buten-1-ol 97+%

Warning

AC36821-0100 AC36821-0500
200035-274EA 200 USD
200035-274 103465-330
3-Bromo-3-buten-1-ol 97+%
3-Bromo-3-buten-1-ol
Formula: C₄H₇BrO
Boiling Pt: 175 °C (760 mmHg)
Density: 1.522 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00154041
CAS Number: 76334-36-6

Order Now

Specification Test Results

Appearance Clear brown Liquid
Infrared spectrum Conforms
Refractive index 1.5000 to 1.5020 (20 °C, 589 nm)
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR