1-(3-Aminopropyl)pyrrolidine 98%
|
Danger
|
Synonyms:
3-(Pyrrolidin-1-yl)propan-1-amine
, N-(3-Aminopropyl)pyrrolidine
AC36809-0250
AC36809-0050
200025-990EA
257
USD
200025-990
200013-466
1-(3-Aminopropyl)pyrrolidine 98%
1-(3-Aminopropyl)pyrrolidine
Formula:
C₇H₁₆N₂ MW: 128.22 g/mol Boiling Pt: 191…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014100 CAS Number: 23159-07-1 EINECS: 245-465-2 UN: 2735 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4700 to 1.4720 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...