3-Amino-5-methyl-1H-pyrazole 97%

Warning

Synonyms: 3-Amino-5-methylpyrazole5-Methyl-1H-pyrazol-3-amine

AC36803-1000 AC36803-0250
200035-234EA 219 USD
200035-234 200009-768
3-Amino-5-methyl-1H-pyrazole 97%
3-Amino-5-methyl-1H-pyrazole
Formula: C₄H₇N₃
MW: 97.12 g/mol
Melting Pt: 41…43 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075180
CAS Number: 31230-17-8

Order Now

Specification Test Results

Appearance Light yellow to orange Solid or viscous liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR