4-Fluoro-2-methylphenylboronic acid 97%

Warning

Synonyms: 2-Methyl-4-fluorophenylboronic acid4-Fluoro-2-methylbenzeneboronic acid

AC36794-0050 AC36794-0010 AC36794-0250
103512-844EA 568.1 USD
103512-844 200041-998 200010-400
4-Fluoro-2-methylphenylboronic acid 97%
4-Fluoro-2-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153.95 g/mol
Melting Pt: 194…196 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093072
CAS Number: 139911-29-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR