3-Carboxyphenylboronic acid pinacol ester 97%
|
Warning
|
Synonyms:
3-Carboxybenzeneboronic acid pinacol ester
, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
AC36757-0010
AC36757-0050
200022-956EA
130
USD
200022-956
200051-910
3-Carboxyphenylboronic acid pinacol ester 97%
3-Carboxyphenylboronic acid pinacol ester
Formula:
C₁₃H₁₇BO₄ MW: 248.083 g/mol Melting Pt: 206…211 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03411930 CAS Number: 269409-73-6 |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 206 to 211 °C |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...