4-Hydroxy-3-methoxyphenylboronic acid pinacol ester 97%

Warning

Synonyms: 2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

AC36754-0050 AC36754-0010
200022-768EA 283 USD
200022-768 200034-870
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester 97%
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₉BO₄
MW: 250.099 g/mol
Melting Pt: 101…106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093723
CAS Number: 269410-22-2

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 101 to 106 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR