Benzeneruthenium(II) chloride dimer 97%

Warning

Synonyms: Dichloro(benzene)ruthenium(II) dimer

AC36748-0010
200030-488EA 132 USD
200030-488
Benzeneruthenium(II) chloride dimer 97%
Benzeneruthenium(II) chloride dimer
Formula: C₁₂H₁₂Cl₄Ru₂
MW: 500.17 g/mol
Melting Pt: 249 °C
Storage Temperature: Ambient
MDL Number: MFCD00064686
CAS Number: 37366-09-9

Order Now

Specification Test Results

Appearance Red to brown Powder or crystals
Infrared spectrum Conforms
Ruthenium (Ru) 39.2 to 41.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR