(R)-(+)-4-Bromo-α-methylbenzylamine hydrochloride 97%, 99+% ee
|
Warning
|
Synonyms:
(R)-1-(4-Bromophenyl)ethylamine Hydrochloride
, (R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride
AC36717-0050
AC36717-0010
AC36717-0250
200039-406EA
64
USD
200039-406
200033-380
103512-792
(R)-(+)-4-Bromo-α-methylbenzylamine hydrochloride 97%, 99+% ee
(R)-(+)-4-Bromo-α-methylbenzylamine hydrochloride
Formula:
C₈H₁₁BrClN MW: 236.54 g/mol Melting Pt: 238…245 °C Storage Temperature: Ambient |
MDL Number:
MFCD02259386 EINECS: 256-245-0 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 238 to 245 °C |
HPLC | ≥96.0 % |
Enantiomeric excess | ≥99.0 % (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...