2-(Piperazin-1-yl)pyrazine 98%

Danger

Synonyms: 1-(2-Pyrazinyl)piperazine

AC36713-0050 AC36713-0010
200004-584EA 56.7 USD
200004-584 200008-322
2-(Piperazin-1-yl)pyrazine 98%
2-(Piperazin-1-yl)pyrazine

Appearance: Yellow Crystalline powder and/or chunks

Formula: C₈H₁₂N₄
MW: 164.212 g/mol
Boiling Pt: 105…108 °C (.5mbar)
Storage Temperature: Ambient
MDL Number: MFCD00040766
CAS Number: 34803-68-4
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR