Triphenylphosphine 1.0 - 1.5 mmol/g on resin, cross-linked with 1% DVB

Warning

AC36687-0050
200033-100EA 85.9 USD
200033-100
Triphenylphosphine 1.0 - 1.5 mmol/g on resin, cross-linked with 1% DVB
Triphenylphosphine
Formula: C₁₈H₁₅P
MW: 262.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20489348
CAS Number: 39319-11-4

Order Now

Specification Test Results

Appearance White to yellow Beads
Particle size 100 to 200 mesh
Crosslinking 1 % DVB
Loading 1 to 1.5 mmol/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR