Cyclopropanecarboxaldehyde 98%

Danger

Synonyms: Cyclopropanecarbaldehyde

AC36682-0050 AC36682-0250 AC36682-0010
200030-820EA 36.8 USD
200030-820 103527-902 200006-168
Cyclopropanecarboxaldehyde 98%
Cyclopropanecarboxaldehyde
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 98…101 °C
Density: 0.938 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00012261
CAS Number: 1489-69-6
UN: 1989
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4285 to 1.4305 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR