4-Hydroxyphenylboronic acid pinacol ester 97%

Warning

Synonyms: 4-Hydroxybenzeneboronic acid pinacol ester4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

AC36642-0050
200005-320EA 215.36 USD
200005-320
4-Hydroxyphenylboronic acid pinacol ester 97%
4-Hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₇BO₃
MW: 220.08 g/mol
Melting Pt: 112…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093756
CAS Number: 269409-70-3

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 112 to 117 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR