α-Bromostyrene 95% stabilized
|
Warning
|
AC36580-0250
AC36580-0050
AC36580-1000
200018-088EA
206
USD
200018-088
103512-780
200063-778
α-Bromostyrene 95% stabilized
α-Bromostyrene
Formula:
C₈H₇Br MW: 183.0452 g/mol Boiling Pt: 67…70 °C (4 mmHg) Melting Pt: –44 °C Density: 1.41 g/cm³ Flash Pt: 86 °C Storage Temperature: Refrigerator |
EINECS:
202-702-4 |
Specification Test Results
Appearance | Clear colorless to orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5870 to 1.5910 (20 °C, 589 nm) |
GC | ≥94 % |
Stabilizer | About 0.05% 3,5-di-tert-Butylcatechol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...