Coconut oil, pure, refined

AC36547-5000 AC36547-1000
200030-762EA 19.8 USD
200030-762 200018-562
Coconut oil, pure, refined
Coconut oil
Melting Pt: 23…26 °C
Flash Pt: >200 °C
Storage Temperature: Refrigerator
EINECS: 232-282-8

Order Now

Specification Test Results

Appearance Colorless to white to light yellow Oily liquid or solid
Acid value ≤0.5 mg KOH/g
Iodine value 7 to 12 g I2/100 g
Peroxide value ≤5 meq O2/kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR