7-Hydroxyflavanone 98%
|
Warning
|
AC36417-0010
200030-202EA
48.8
USD
200030-202
7-Hydroxyflavanone 98%
7-Hydroxyflavanone
Specification Test Results
Appearance | Light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 189 to 197 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...