1,1'-Bis(diisopropylphosphino)ferrocene 98%
|
Warning
|
Synonyms:
1,1'-Bis(di-i-propylphosphino)ferrocene
, DiPPF
AC36387-0025
103512-738EA
327.38
USD
103512-738
1,1'-Bis(diisopropylphosphino)ferrocene 98%
1,1'-Bis(diisopropylphosphino)ferrocene
Formula:
C₂₂H₂₈FeP₂ MW: 410,25 g/mol Melting Pt: 53…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684559 CAS Number: 97239-80-0 |
Specification Test Results
Appearance | Yellow to orange Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 49 to 53 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...